2-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049978-500MG
049978-2.5G
101889-812EA
364.62
USD
101889-812
101889-814
2-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₉ClF₃NO |
MDL Number:
MFCD13560380 CAS Number: 1219982-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...