3-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049977-500MG 049977-2.5G
101889-808EA 364.62 USD
101889-808 101889-810
3-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClF₃NO
MDL Number: MFCD13560379
CAS Number: 1219960-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR