4-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049976-500MG 049976-2.5G
101889-804EA 364.62 USD
101889-804 101889-806
4-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[2-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClF₃NO
MDL Number: MFCD13560378
CAS Number: 1219960-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR