3-[([1,1'-Biphenyl]-2-yloxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049975-500MG
049975-2.5G
101889-800EA
364.62
USD
101889-800
101889-802
3-[([1,1'-Biphenyl]-2-yloxy)methyl]pyrrolidine hydrochloride
3-[([1,1'-Biphenyl]-2-yloxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₀ClNO |
MDL Number:
MFCD13560377 CAS Number: 1220028-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...