2-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049974-500MG
049974-2.5G
101889-796EA
186.15
USD
101889-796
101889-798
2-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride
2-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₉H₂₄ClNO |
MDL Number:
MFCD13560376 CAS Number: 1220031-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...