3-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049973-500MG 049973-2.5G
101889-792EA 364.62 USD
101889-792 101889-794
3-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride
3-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD13560375
CAS Number: 1220019-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR