4-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049972-500MG 049972-2.5G
101889-788EA 364.62 USD
101889-788 101889-790
4-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride
4-[2-([1,1'-Biphenyl]-2-yloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD13560374
CAS Number: 1220037-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR