3-[(2-Ethoxyphenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049971-500MG
049971-2.5G
101889-784EA
364.62
USD
101889-784
101889-786
3-[(2-Ethoxyphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Ethoxyphenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀ClNO₂ |
MDL Number:
MFCD13560373 CAS Number: 1219949-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...