3-[(2-Methoxyphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049967-500MG 049967-2.5G
101889-768EA 364.62 USD
101889-768 101889-770
3-[(2-Methoxyphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Methoxyphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO₂
MDL Number: MFCD13560369
CAS Number: 51535-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR