2-Methoxyphenyl-2-(4-piperidinyl)ethyl ether hydrochloride
Supplier: Matrix Scientific
049964-500MG
049964-2.5G
101889-756EA
364.62
USD
101889-756
101889-758
2-Methoxyphenyl-2-(4-piperidinyl)ethyl ether hydrochloride
2-Methoxyphenyl-2-(4-piperidinyl)ethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO₂ |
MDL Number:
MFCD13560366 CAS Number: 614731-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...