2-Chloro-5-methylphenyl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

049963-500MG 049963-2.5G
101889-752EA 364.62 USD
101889-752 101889-754
2-Chloro-5-methylphenyl-3-pyrrolidinylmethyl ether hydrochloride
2-Chloro-5-methylphenyl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560365
CAS Number: 1220019-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR