2-[2-(2-Chloro-5-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049962-500MG 049962-2.5G
101889-748EA 364.62 USD
101889-748 101889-750
2-[2-(2-Chloro-5-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Chloro-5-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560364
CAS Number: 1219972-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR