2-[2-(2,5-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049958-500MG 049958-2.5G
101889-732EA 364.62 USD
101889-732 101889-734
2-[2-(2,5-Dichlorophenoxy)ethyl]piperidine hydrochloride
2-[2-(2,5-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560360
CAS Number: 1219981-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR