3-[2-(2,5-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049957-500MG 049957-2.5G
101889-728EA 364.62 USD
101889-728 101889-730
3-[2-(2,5-Dichlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2,5-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560359
CAS Number: 1220036-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR