2-(2-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
Supplier: Matrix Scientific
049954-500MG
049954-2.5G
101889-716EA
364.62
USD
101889-716
101889-718
2-(2-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
2-(2-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₇Cl₄NO MW: 345.1 g/mol |
MDL Number:
MFCD13560356 CAS Number: 1219972-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...