2-(3-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
Supplier: Matrix Scientific
049953-500MG
049953-2.5G
101889-712EA
364.62
USD
101889-712
101889-714
2-(3-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
2-(3-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₇Cl₄NO |
MDL Number:
MFCD13560355 CAS Number: 1219949-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...