2-(4-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride

Supplier: Matrix Scientific

049952-500MG 049952-2.5G
101889-708EA 364.62 USD
101889-708 101889-710
2-(4-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride
2-(4-Piperidinyl)ethyl-2,4,5-trichlorophenylether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇Cl₄NO
MDL Number: MFCD13560354
CAS Number: 1220036-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR