3-[(2,4-Dichlorophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049951-500MG 049951-2.5G
101889-704EA 364.62 USD
101889-704 101889-706
3-[(2,4-Dichlorophenoxy)methyl]pyrrolidine hydrochloride
3-[(2,4-Dichlorophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₃NO
MW: 282.6 g/mol
MDL Number: MFCD13560353
CAS Number: 1219949-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR