2-[2-(2,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049950-500MG 049950-2.5G
101889-700EA 186.15 USD
101889-700 101889-702
2-[2-(2,4-Dichlorophenoxy)ethyl]piperidine hydrochloride
2-[2-(2,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560352
CAS Number: 1219972-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR