2-[2-(2-Chloro-4-fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049946-500MG 049946-2.5G
101889-684EA 364.62 USD
101889-684 101889-686
2-[2-(2-Chloro-4-fluorophenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Chloro-4-fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂FNO
MDL Number: MFCD13560348
CAS Number: 1219981-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR