3-[(4-Bromo-2-chlorophenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049943-500MG
049943-2.5G
101889-672EA
364.62
USD
101889-672
101889-674
3-[(4-Bromo-2-chlorophenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Bromo-2-chlorophenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄BrCl₂NO |
MDL Number:
MFCD13560345 CAS Number: 1219949-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...