2-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049942-500MG
049942-2.5G
101889-668EA
364.62
USD
101889-668
101889-670
2-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈BrCl₂NO |
MDL Number:
MFCD13560344 CAS Number: 1219957-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...