3-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049941-500MG 049941-2.5G
101889-664EA 364.62 USD
101889-664 101889-666
3-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrCl₂NO
MDL Number: MFCD13560343
CAS Number: 1220018-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR