4-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049940-500MG 049940-2.5G
101889-660EA 364.62 USD
101889-660 101889-662
4-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromo-2-chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrCl₂NO
MDL Number: MFCD13560342
CAS Number: 1219981-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR