3-[(2,3-Dichlorophenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049939-500MG
049939-2.5G
101889-656EA
364.62
USD
101889-656
101889-658
3-[(2,3-Dichlorophenoxy)methyl]pyrrolidine hydrochloride
3-[(2,3-Dichlorophenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄Cl₃NO |
MDL Number:
MFCD13560341 CAS Number: 1219949-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...