3-[2-(2,3-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049937-500MG 049937-2.5G
101889-648EA 364.62 USD
101889-648 101889-650
3-[2-(2,3-Dichlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2,3-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560339
CAS Number: 1219972-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR