4-[2-(2,3-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049936-500MG 049936-2.5G
101889-644EA 364.62 USD
101889-644 101889-646
4-[2-(2,3-Dichlorophenoxy)ethyl]piperidine hydrochloride
4-[2-(2,3-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560338
CAS Number: 1219981-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR