3-[(2-Chlorophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049935-500MG 049935-2.5G
101889-640EA 364.62 USD
101889-640 101889-642
3-[(2-Chlorophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Chlorophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅Cl₂NO
MDL Number: MFCD13560337
CAS Number: 1018446-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR