3-[2-(2-Chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049933-500MG 049933-2.5G
101889-632EA 364.62 USD
101889-632 101889-634
3-[2-(2-Chlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560335
CAS Number: 1018446-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR