4-[2-(2-Chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049932-500MG 049932-2.5G
101889-628EA 364.62 USD
101889-628 101889-630
4-[2-(2-Chlorophenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560334
CAS Number: 1219957-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR