2-[2-(2-Fluorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049930-500MG 049930-2.5G
101889-620EA 364.62 USD
101889-620 101889-622
2-[2-(2-Fluorophenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Fluorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClFNO
MDL Number: MFCD13560332
CAS Number: 1220018-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR