3-[(2-Bromo-4-methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049927-500MG 049927-2.5G
101889-608EA 364.62 USD
101889-608 101889-610
3-[(2-Bromo-4-methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Bromo-4-methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD13560329
CAS Number: 1219980-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR