3-[2-(2-Bromo-4-methylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049925-500MG
049925-2.5G
101889-600EA
364.62
USD
101889-600
101889-602
3-[2-(2-Bromo-4-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Bromo-4-methylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁BrClNO |
MDL Number:
MFCD13560327 CAS Number: 1219957-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...