4-[2-(2-Bromo-4-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049924-500MG 049924-2.5G
101889-596EA 364.62 USD
101889-596 101889-598
4-[2-(2-Bromo-4-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Bromo-4-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560326
CAS Number: 1219972-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR