2-[2-(2,4-Dibromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049922-500MG 049922-2.5G
101889-588EA 364.62 USD
101889-588 101889-590
2-[2-(2,4-Dibromophenoxy)ethyl]piperidine hydrochloride
2-[2-(2,4-Dibromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Br₂ClNO
MDL Number: MFCD13560324
CAS Number: 1220028-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR