4-[2-(2,4-Dibromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049920-500MG 049920-2.5G
101889-580EA 364.62 USD
101889-580 101889-582
4-[2-(2,4-Dibromophenoxy)ethyl]piperidine hydrochloride
4-[2-(2,4-Dibromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Br₂ClNO
MDL Number: MFCD13560322
CAS Number: 1219972-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR