3-[(2-Bromophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049919-500MG 049919-2.5G
101889-576EA 364.62 USD
101889-576 101889-578
3-[(2-Bromophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Bromophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD13560321
CAS Number: 1220036-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR