2-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049918-500MG 049918-2.5G
101889-572EA 364.62 USD
101889-572 101889-574
2-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560320
CAS Number: 1219972-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR