3-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049917-500MG 049917-2.5G
101889-568EA 364.62 USD
101889-568 101889-570
3-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560319
CAS Number: 1219957-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR