4-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049916-500MG 049916-2.5G
101889-564EA 364.62 USD
101889-564 101889-566
4-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560318
CAS Number: 1220018-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR