2-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049914-500MG 049914-2.5G
101889-556EA 364.62 USD
101889-556 101889-558
2-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride
2-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO₂
MDL Number: MFCD13560316
CAS Number: 1220028-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR