3-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049913-500MG 049913-2.5G
101889-552EA 364.62 USD
101889-552 101889-554
3-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride
3-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO₂
MDL Number: MFCD13560315
CAS Number: 1219981-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR