3-[(3-Methoxypropoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049911-500MG 049911-2.5G
101889-544EA 364.62 USD
101889-544 101889-546
3-[(3-Methoxypropoxy)methyl]pyrrolidine hydrochloride
3-[(3-Methoxypropoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₉H₂₀ClNO₂
MDL Number: MFCD13560313
CAS Number: 1220031-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR