3-{[2-(Allyloxy)ethoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049907-500MG 049907-2.5G
101889-528EA 364.62 USD
101889-528 101889-530
3-{[2-(Allyloxy)ethoxy]methyl}pyrrolidine hydrochloride
3-{[2-(Allyloxy)ethoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD13560309
CAS Number: 1219971-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR