2-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049906-500MG 049906-2.5G
101889-524EA 364.62 USD
101889-524 101889-526
2-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride
2-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO₂
MDL Number: MFCD13560308
CAS Number: 1220018-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR