3-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049905-500MG
049905-2.5G
101889-520EA
364.62
USD
101889-520
101889-522
3-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride
3-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₄ClNO₂ |
MDL Number:
MFCD13560307 CAS Number: 1219949-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...