4-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049904-500MG 049904-2.5G
101889-516EA 364.62 USD
101889-516 101889-518
4-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride
4-{2-[2-(Allyloxy)ethoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO₂
MDL Number: MFCD13560306
CAS Number: 1219971-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR