2-Methyl-2-propenyl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

049903-500MG 049903-2.5G
101889-512EA 364.62 USD
101889-512 101889-514
2-Methyl-2-propenyl-3-pyrrolidinylmethyl ether hydrochloride
2-Methyl-2-propenyl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO
MDL Number: MFCD13560305
CAS Number: 1219981-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR