2-{2-[(2-Methyl-2-propenyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049902-500MG 049902-2.5G
101889-508EA 364.62 USD
101889-508 101889-510
2-{2-[(2-Methyl-2-propenyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2-Methyl-2-propenyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO
MDL Number: MFCD13560304
CAS Number: 1219949-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR