3-{2-[(2-Methyl-2-propenyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049901-500MG 049901-2.5G
101889-504EA 364.62 USD
101889-504 101889-506
3-{2-[(2-Methyl-2-propenyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(2-Methyl-2-propenyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClNO
MDL Number: MFCD13560303
CAS Number: 1220018-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR